missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Cellulose, FCC, 44-50%, Spectrum™ Chemical
E1036, 9004-57-3
$571.54 - $4500.75
Chemical Identifiers
| CAS | 9004-57-3 |
|---|---|
| Molecular Formula | C23H24N6O4 |
| Molecular Weight (g/mol) | 448.48 |
| InChI Key | ARSKJXYLLONUAJ-UHFFFAOYSA-N |
| IUPAC Name | 2-[butyl({4-[2-(2,6-dicyano-4-nitrophenyl)diazen-1-yl]-3-methylphenyl})amino]ethyl acetate |
| SMILES | CCCCN(CCOC(C)=O)C1=CC(C)=C(C=C1)N=NC1=C(C=C(C=C1C#N)[N+]([O-])=O)C#N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18602533
|
Spectrum Chemical Mfg Cor
E1036125GM |
125 g | Poly Bottle |
Each for $571.54
|
|
||||
|
18602534
|
Spectrum Chemical Mfg Cor
E1036500GM |
500 g | Poly Bottle |
Each for $1,377.89
|
|
||||
|
18602535
|
Spectrum Chemical Mfg Cor
E103625KG |
2.5 kg | Poly Pail |
Each for $4,500.75
|
|
||||
Description
Spectrum™ Chemical Ethyl Cellulose, FCC , a derivative of cellulose, is often used as an emulsifier food additive. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 9004-57-3 | |
| 448.48 | |
| 2-[butyl({4-[2-(2,6-dicyano-4-nitrophenyl)diazen-1-yl]-3-methylphenyl})amino]ethyl acetate |
| C23H24N6O4 | |
| ARSKJXYLLONUAJ-UHFFFAOYSA-N | |
| CCCCN(CCOC(C)=O)C1=CC(C)=C(C=C1)N=NC1=C(C=C(C=C1C#N)[N+]([O-])=O)C#N |
Specifications
| 9004-57-3 | |
| C23H24N6O4 | |
| ARSKJXYLLONUAJ-UHFFFAOYSA-N | |
| 2-[butyl({4-[2-(2,6-dicyano-4-nitrophenyl)diazen-1-yl]-3-methylphenyl})amino]ethyl acetate | |
| 44 to 50% | |
| 0.5% | |
| Poly Bottle |
| 100% | |
| 125 g | |
| CCCCN(CCOC(C)=O)C1=CC(C)=C(C=C1)N=NC1=C(C=C(C=C1C#N)[N+]([O-])=O)C#N | |
| 448.48 | |
| FCC | |
| 3% |