missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Cellulose, FCC, 44-50%, Spectrum™ Chemical

Catalog No. 18602533
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602533 125 g Poly Bottle
18602534 500 g Poly Bottle
18602535 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18602533 Supplier Spectrum Chemical Mfg Cor Supplier No. E1036125GM

E1036, 9004-57-3

Spectrum™ Chemical Ethyl Cellulose, FCC , a derivative of cellulose, is often used as an emulsifier food additive. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9004-57-3
Molecular Formula C23H24N6O4
Molecular Weight (g/mol) 448.48
InChI Key ARSKJXYLLONUAJ-UHFFFAOYSA-N
IUPAC Name 2-[butyl({4-[2-(2,6-dicyano-4-nitrophenyl)diazen-1-yl]-3-methylphenyl})amino]ethyl acetate
SMILES CCCCN(CCOC(C)=O)C1=CC(C)=C(C=C1)N=NC1=C(C=C(C=C1C#N)[N+]([O-])=O)C#N

Specifications

CAS Max % 100%
Quantity 125 g
Percent Purity 44 to 50%
Grade FCC
Ignition Residue 0.5%
Loss on Drying 3%
Packaging Poly Bottle