missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Carbazate 99.0+%, TCI America™
Supplier: TCI America C109725G
Specifications
| Ethyl Carbazate | |
| 47°C | |
| 118°C | |
| MFCD00007595 | |
| 2811 | |
| VYSYZMNJHYOXGN-UHFFFAOYSA-N | |
| ethyl N-aminocarbamate | |
| 20064 | |
| ≥99.0% (T) |
| 4114-31-2 | |
| White | |
| C3H8N2O2 | |
| 25 g | |
| ethyl carbazate, ethyl hydrazinecarboxylate, ethoxycarbohydrazide, hydrazinecarboxylic acid, ethyl ester, carbethoxyhydrazine, ethylcarbazate, ethyl carbazinate, carboethoxyhydrazine, monocarbethoxyhydrazine, n-carbethoxy hydrazine | |
| CCOC(=O)NN | |
| 104.109 | |
| 104.11 | |
| Crystalline Powder |
Chemical Identifiers
| 4114-31-2 | |
| 104.109 | |
| VYSYZMNJHYOXGN-UHFFFAOYSA-N | |
| 20064 | |
| CCOC(=O)NN |
| C3H8N2O2 | |
| MFCD00007595 | |
| ethyl carbazate, ethyl hydrazinecarboxylate, ethoxycarbohydrazide, hydrazinecarboxylic acid, ethyl ester, carbethoxyhydrazine, ethylcarbazate, ethyl carbazinate, carboethoxyhydrazine, monocarbethoxyhydrazine, n-carbethoxy hydrazine | |
| ethyl N-aminocarbamate |
Safety and Handling
RTECSNumber : FE2545000
TSCA : Yes