missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Carbazate 99.0+%, TCI America™

Catalog No. p-7124790
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C109725G 25 g
C1097500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C109725G Supplier TCI America Supplier No. C109725G
Only null left

Chemical Identifiers

CAS 4114-31-2
Molecular Formula C3H8N2O2
Molecular Weight (g/mol) 104.109
MDL Number MFCD00007595
InChI Key VYSYZMNJHYOXGN-UHFFFAOYSA-N
Synonym ethyl carbazate, ethyl hydrazinecarboxylate, ethoxycarbohydrazide, hydrazinecarboxylic acid, ethyl ester, carbethoxyhydrazine, ethylcarbazate, ethyl carbazinate, carboethoxyhydrazine, monocarbethoxyhydrazine, n-carbethoxy hydrazine
PubChem CID 20064
IUPAC Name ethyl N-aminocarbamate
SMILES CCOC(=O)NN

Specifications

Melting Point 47°C
Color White
Boiling Point 118°C
Quantity 25 g
UN Number 2811
Formula Weight 104.11
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl Carbazate
RTECSNumber FE2545000
TSCA Yes