missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Carbamate 98.0+%, TCI America™

Catalog No. C0028500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0028500G 500 g
C002825G 25 g
C0028100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C0028500G Supplier TCI America Supplier No. C0028500G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 51-79-6
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.094
MDL Number MFCD00007966
InChI Key JOYRKODLDBILNP-UHFFFAOYSA-N
Synonym urethane, urethan, ethylurethane, carbamic acid ethyl ester, ethyl urethane, carbamic acid, ethyl ester, ethylcarbamate, pracarbamine, leucethane, pracarbamin
PubChem CID 5641
ChEBI CHEBI:17967
IUPAC Name ethyl carbamate
SMILES CCOC(=O)N

Specifications

Melting Point 50°C
Color White
Boiling Point 184°C
Quantity 500 g
Formula Weight 89.09
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl Carbamate
EINECSNumber (2)-1712
RTECSNumber FA8400000
TSCA Yes