accessibility menu, dialog, popup

UserName

CH3ONH2O

CAS RN 51-79-6

IUPAC Name: ethyl carbamate
Synonyms: urethane pracarbamine O-ethyl carbamate (urethane) pracarbamin U-compound ethylester kyseliny karbaminove O-ethylurethane uretan etylowy urethan (substance) ethyl urethane
Molecular Weight (g/mol): 89.09
Molecular Formula: C3H7NO2
InChi Key: JOYRKODLDBILNP-UHFFFAOYSA-N
SMILES: CCOC(N)=O
Boiling Point 
  • (2)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (3)
  • (3)
  • (2)
Physical Form 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)