missing translation for 'onlineSavingsMsg'
Learn More

Ethyl carbamate, 98%

Catalog No. p-7059534
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4480418 50 g
AA4480430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4480418 Supplier Thermo Scientific Chemicals Supplier No. 04480418

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 51-79-6 | C3H7NO2 | 89.094 g/mol

Ethyl carbamate in its molten state is good solvent for various organic materials, preparation & modification of amino resins, as solubilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51-79-6
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.094
MDL Number MFCD00007966
InChI Key JOYRKODLDBILNP-UHFFFAOYSA-N
Synonym urethane, urethan, ethylurethane, carbamic acid ethyl ester, ethyl urethane, carbamic acid, ethyl ester, ethylcarbamate, pracarbamine, leucethane, pracarbamin
PubChem CID 5641
ChEBI CHEBI:17967
IUPAC Name ethyl carbamate
SMILES CCOC(=O)N

Specifications

Melting Point 48°C to 50°C
Color White
Boiling Point 182°C to 184°C
Flash Point >92°C (198°F)
Linear Formula H2NCO2C2H5
Quantity 50 g
Beilstein 635810
Merck Index 14,9877
Solubility Information Soluble in chloroform.
Formula Weight 89.09
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material Ethyl carbamate
Show More Show Less
Hazard Category H302-H350
Hazard Statement GHS H Statement
H350
May cause cancer.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
EINECSNumber 200-123-1
RTECSNumber FA8400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only