missing translation for 'onlineSavingsMsg'
Learn More

Ethyl bromofluoroacetate, 97%

Catalog No. p-7025867
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2157903 1 g
AAB2157906 5 g
AAB2157914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2157903 Supplier Thermo Scientific Chemicals Supplier No. B2157903

CAS: 401-55-8 | C4H6BrFO2 | 184.992 g/mol

Ethyl bromofluoroacetate is used as an intermediate useful in the synthesis of polyfluoroalkyl-substituted fluoro enol ethers and N-protected α-fluoroglycines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl bromofluoroacetate is used as an intermediate useful in the synthesis of polyfluoroalkyl-substituted fluoro enol ethers and N-protected α-fluoroglycines.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.

Chemical Identifiers

CAS 401-55-8
Molecular Formula C4H6BrFO2
Molecular Weight (g/mol) 184.992
MDL Number MFCD00042095
InChI Key ULNDTPIRBQGESN-UHFFFAOYSA-N
Synonym ethyl bromofluoroacetate, bromofluoroacetic acid ethyl ester, ethyl-2-bromofluoroacetate, ethylbromofluoroacetate, acetic acid, bromofluoro-, ethyl ester, bromofluoroacetate, ethyl fluorobromoacetate, acmc-209jb9, ksc236i3t
PubChem CID 2733407
IUPAC Name ethyl 2-bromo-2-fluoroacetate
SMILES CCOC(=O)C(F)Br

Specifications

Density 1.54
Boiling Point 154°C
Flash Point 70°C (158°F)
Refractive Index 1.4255
Quantity 1 g
UN Number UN2810
Solubility Information It is soluble in water.
Formula Weight 184.99
Percent Purity 97%
Chemical Name or Material Ethyl bromofluoroacetate
Hazard Category H227-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H331-H318-H227-H315-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes serious eye damage.
Combustible liquid.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 206-928-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only