Learn More
Ethyl bromodifluoroacetate, 97%
CAS: 667-27-6 | C4H5BrF2O2 | 202.983 g/mol
$127.63 - $127.63
Chemical Identifiers
| CAS | 667-27-6 |
|---|---|
| Molecular Formula | C4H5BrF2O2 |
| Molecular Weight (g/mol) | 202.983 |
| MDL Number | MFCD00042069 |
| InChI Key | IRSJDVYTJUCXRV-UHFFFAOYSA-N |
| Synonym | ethyl bromodifluoroacetate, ethylbromodifluoroacetate, bromodifluoroacetic acid ethyl ester, acetic acid, bromodifluoro-, ethyl ester, ethyl difluorobromoacetate, ethyl 2-bromo-2,2-difluoro-acetate, 2-bromo-2,2-difluoro-1-ethoxyethan-1-one, 2-bromo-2,2-difluoro-1-ethoxy-1-oxoethane, ethylbrom difluor acetat, pubchem16953 |
| PubChem CID | 69585 |
| IUPAC Name | ethyl 2-bromo-2,2-difluoroacetate |
| SMILES | CCOC(=O)C(F)(F)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2046106
|
Thermo Scientific Chemicals
B2046106 |
5 g |
Each for $127.63
|
|
|||||
Description
Ethyl bromodifluoroacetate, is used for the treatment of the Reformat sky reagent with aldehydes and ketones affords 2,2-difluoro-3-hydroxy esters. It is also used in the analysis of reagent purity through IR, NMR.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl bromodifluoroacetate, is used for the treatment of the Reformat sky reagent with aldehydes and ketones affords 2,2-difluoro-3-hydroxy esters. It is also used in the analysis of reagent purity through IR, NMR.
Solubility
Soluble in most organic solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Chemical Identifiers
| 667-27-6 | |
| 202.983 | |
| IRSJDVYTJUCXRV-UHFFFAOYSA-N | |
| 69585 | |
| CCOC(=O)C(F)(F)Br |
| C4H5BrF2O2 | |
| MFCD00042069 | |
| ethyl bromodifluoroacetate, ethylbromodifluoroacetate, bromodifluoroacetic acid ethyl ester, acetic acid, bromodifluoro-, ethyl ester, ethyl difluorobromoacetate, ethyl 2-bromo-2,2-difluoro-acetate, 2-bromo-2,2-difluoro-1-ethoxyethan-1-one, 2-bromo-2,2-difluoro-1-ethoxy-1-oxoethane, ethylbrom difluor acetat, pubchem16953 | |
| ethyl 2-bromo-2,2-difluoroacetate |
Specifications
| 667-27-6 | |
| 112°C to 114°C | |
| C4H5BrF2O2 | |
| MFCD00042069 | |
| UN2924 | |
| ethyl bromodifluoroacetate, ethylbromodifluoroacetate, bromodifluoroacetic acid ethyl ester, acetic acid, bromodifluoro-, ethyl ester, ethyl difluorobromoacetate, ethyl 2-bromo-2,2-difluoro-acetate, 2-bromo-2,2-difluoro-1-ethoxyethan-1-one, 2-bromo-2,2-difluoro-1-ethoxy-1-oxoethane, ethylbrom difluor acetat, pubchem16953 | |
| IRSJDVYTJUCXRV-UHFFFAOYSA-N | |
| ethyl 2-bromo-2,2-difluoroacetate | |
| 69585 | |
| 97% |
| 1.583 | |
| 21°C (69°F) | |
| 1.388 | |
| 5 g | |
| 1906095 | |
| Soluble in most organic solvents. | |
| CCOC(=O)C(F)(F)Br | |
| 202.983 | |
| 202.99 | |
| Ethyl bromodifluoroacetate |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber : 211-567-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only