missing translation for 'onlineSavingsMsg'
Learn More

Ethyl bromoacetate, 98%

Catalog No. p-127436
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC158591000 100 g Glass bottle
AC158595000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC158591000 Supplier Thermo Scientific Chemicals Supplier No. 158591000

CAS: 105-36-2 | C4H7BrO2 | 167 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 105-36-2
MDL Number MFCD00000191
InChI Key PQJJJMRNHATNKG-UHFFFAOYSA-N
Synonym ethyl bromoacetate, antol, ethyl bromacetate, bromoacetic acid ethyl ester, ethyl monobromoacetate, acetic acid, bromo-, ethyl ester, ethoxycarbonylmethyl bromide, bromoacetic acid, ethyl ester, ethyl alpha-bromoacetate, ethyl bromo acetate
PubChem CID 7748
IUPAC Name ethyl 2-bromoacetate
SMILES CCOC(=O)CBr

Specifications

Melting Point -38.0°C
Density 1.5060g/mL
Boiling Point 159.0°C
Flash Point 47°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C4H7BrO2
Refractive Index 1.4500 to 1.4520
Linear Formula BrCH2CO2C2H5
Quantity 100 g
Beilstein 02, 214
Fieser 02,192; 13,60
Specific Gravity 1.506
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in alcohol, benzene and ether
Molecular Weight (g/mol) 167
Formula Weight 167
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl bromoacetate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if inhaled.
Fatal if swallowed.
Fatal in contact with skin.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Keep away from heat/sparks/open flames/hot
EINECSNumber 203-290-9
RTECSNumber AF6000000
TSCA TSCA

RUO – Research Use Only