accessibility menu, dialog, popup

UserName

CAS RN 105-36-2

IUPAC Name: ethyl 2-bromoacetate
Synonyms: ethyl bromoacetate
Molecular Weight (g/mol): 167
Molecular Formula: C4H7BrO2
InChi Key: PQJJJMRNHATNKG-UHFFFAOYSA-N
SMILES: CCOC(=O)CBr
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results