missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Bis(2,4-dinitrophenyl)acetate 98.0+%, TCI America™

Catalog No. B08175G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B08175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B08175G Supplier TCI America Supplier No. B08175G

Chemical Identifiers

CAS 5833-18-1
Molecular Formula C16H12N4O10
Molecular Weight (g/mol) 420.29
MDL Number MFCD00051351
InChI Key CMVIALRRHVVWIW-UHFFFAOYSA-N
Synonym ethyl bis 2,4-dinitrophenyl acetate, ethyl 2,2-bis 2,4-dinitrophenyl acetate, bis 2,4-dinitrophenyl acetic acid ethyl ester, ethyl-bis 2,4-dinitrophenyl acetate, acetic acid, bis 2,4-dinitrophenyl-, ethyl ester, benzeneacetic acid, alpha-2,4-dinitrophenyl-2,4-dinitro-, ethyl ester, benzeneacetic acid, .alpha.-2,4-dinitrophenyl-2,4-dinitro-, ethyl ester, acmc-1azlt, 3-09-00-03315 beilstein handbook reference, ethylbis 2,4-dinitrophenyl acetate
PubChem CID 22085
IUPAC Name ethyl 2,2-bis(2,4-dinitrophenyl)acetate
SMILES CCOC(=O)C(C1=C(C=C(C=C1)[N+](=O)[O-])[N+](=O)[O-])C2=C(C=C(C=C2)[N+](=O)[O-])[N+](=O)[O-]

Specifications

Melting Point 156°C
Color Yellow
Quantity 5 g
UN Number 1325
Formula Weight 420.29
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl Bis(2,4-dinitrophenyl)acetate
RTECSNumber AF5650000
TSCA Yes