missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Benzoylformate 96.0+%, TCI America™

Catalog No. p-7118359
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B01095ML 5 mL
B010925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B01095ML Supplier TCI America Supplier No. B01095ML

Chemical Identifiers

CAS 1603-79-8
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
MDL Number MFCD00009120
InChI Key QKLCQKPAECHXCQ-UHFFFAOYSA-N
Synonym ethyl benzoylformate, ethyl phenylglyoxylate, ethyl oxophenylacetate, ethylbenzoylformate, phenylglyoxylic acid ethyl ester, ethyl oxo phenyl acetate, benzeneacetic acid, .alpha.-oxo-, ethyl ester, benzoylformic acid ethyl ester, ethyl benzoyl formate, glyoxylic acid, phenyl-, ethyl ester
PubChem CID 15349
ChEBI CHEBI:84260
IUPAC Name ethyl 2-oxo-2-phenylacetate
SMILES CCOC(=O)C(=O)C1=CC=CC=C1

Specifications

Color Yellow
Quantity 5 mL
Formula Weight 178.19
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Benzoylformate