missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Benzoate 99.0+%, TCI America™

Catalog No. p-7118332
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B006925ML 25 mL
B0069500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B006925ML Supplier TCI America Supplier No. B006925ML
Only null left

Chemical Identifiers

CAS 93-89-0
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00009109
InChI Key MTZQAGJQAFMTAQ-UHFFFAOYSA-N
Synonym benzoic acid, ethyl ester, benzoic ether, ethyl benzenecarboxylate, ethylbenzoate, benzoyl ethyl ether, benzoic acid ethyl ester, ethylester kyseliny benzoove, natural, unii-j115brj15h, fema no. 2422
PubChem CID 7165
IUPAC Name ethyl benzoate
SMILES CCOC(=O)C1=CC=CC=C1

Specifications

Melting Point -34°C
Color Colorless
Boiling Point 212°C
Quantity 25 mL
Formula Weight 150.18
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Benzoate
EINECSNumber (3)-1356
RTECSNumber DH0200000
TSCA Yes