missing translation for 'onlineSavingsMsg'
Learn More

Ethyl benzoate, 99%

Catalog No. AAA1305736
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1305736 500 g
AAA1305722 100 g
AAA130570E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1305736 Supplier Thermo Scientific Chemicals Supplier No. A1305736
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 93-89-0 | C9H10O2 | 150.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Ethyl benzoate is used as a perfume scent. It acts as a food flavoring agent. It is an active component of artificial fruit flavors. Further, it is used in cosmetics and personal care products as fragrance ingredients and preservatives.

Solubility
Miscible with methanol and diethyl ether. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 93-89-0
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00009109
InChI Key MTZQAGJQAFMTAQ-UHFFFAOYSA-N
Synonym benzoic acid, ethyl ester, benzoic ether, ethyl benzenecarboxylate, ethylbenzoate, benzoyl ethyl ether, benzoic acid ethyl ester, ethylester kyseliny benzoove, natural, unii-j115brj15h, fema no. 2422
PubChem CID 7165
IUPAC Name ethyl benzoate
SMILES CCOC(=O)C1=CC=CC=C1

Specifications

Melting Point -34°C
Density 1.049
Boiling Point 212°C to 214°C
Flash Point 84°C (183°F)
Refractive Index 1.505
Quantity 500 g
Beilstein 1908172
Merck Index 14,3766
Solubility Information Miscible with methanol and diethyl ether. Immiscible with water.
Formula Weight 150.18
Percent Purity 99%
Chemical Name or Material Ethyl benzoate
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 202-284-3
RTECSNumber DH0200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only