Learn More
Ethyl Benzoate, 99+%
CAS: 93-89-0 | C9H10O2 | 150.18 g/mol
$80.03 - $223.05
Chemical Identifiers
| CAS | 93-89-0 |
|---|---|
| Molecular Formula | C9H10O2 |
| Molecular Weight (g/mol) | 150.18 |
| MDL Number | MFCD00009109 |
| InChI Key | MTZQAGJQAFMTAQ-UHFFFAOYSA-N |
| Synonym | benzoic acid, ethyl ester, benzoic ether, ethyl benzenecarboxylate, ethylbenzoate, benzoyl ethyl ether, benzoic acid ethyl ester, ethylester kyseliny benzoove, natural, unii-j115brj15h, fema no. 2422 |
| PubChem CID | 7165 |
| IUPAC Name | ethyl benzoate |
| SMILES | CCOC(=O)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC118095000
|
Thermo Scientific Chemicals
118095000 |
500 g | Glass bottle |
Each for $80.03
|
|
||||
|
AC118090025
|
Thermo Scientific Chemicals
118090025 |
2.5 kg | Glass bottle |
Each for $223.05
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 93-89-0 | |
| 150.18 | |
| MTZQAGJQAFMTAQ-UHFFFAOYSA-N | |
| 7165 | |
| CCOC(=O)C1=CC=CC=C1 |
| C9H10O2 | |
| MFCD00009109 | |
| benzoic acid, ethyl ester, benzoic ether, ethyl benzenecarboxylate, ethylbenzoate, benzoyl ethyl ether, benzoic acid ethyl ester, ethylester kyseliny benzoove, natural, unii-j115brj15h, fema no. 2422 | |
| ethyl benzoate |
Specifications
| 93-89-0 | |
| -34.0°C | |
| 1.0450g/mL | |
| 88°C | |
| 99% min. (GC) | |
| C9H10O2 | |
| C6H5CO2C2H5 | |
| 500 g | |
| 1.045 | |
| benzoic acid, ethyl ester, benzoic ether, ethyl benzenecarboxylate, ethylbenzoate, benzoyl ethyl ether, benzoic acid ethyl ester, ethylester kyseliny benzoove, natural, unii-j115brj15h, fema no. 2422 | |
| MTZQAGJQAFMTAQ-UHFFFAOYSA-N | |
| ethyl benzoate | |
| 7165 | |
| 99+% | |
| Ethyl benzoate, 99+% |
| 1mg KOH/g max. | |
| Colorless to Yellow | |
| 212.0°C | |
| Authentic | |
| Glass bottle | |
| 1.5040 to 1.5060 | |
| MFCD00009109 | |
| 09, IV, 285 | |
| 15, 3821 | |
| Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether and chloroform | |
| CCOC(=O)C1=CC=CC=C1 | |
| 150.18 | |
| 150.18 | |
| Liquid |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
EINECSNumber : 202-284-3
RTECSNumber : DH0200000
TSCA : TSCA
RUO – Research Use Only