missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Azidoacetate 95.0+%, TCI America™
Supplier: TCI America E125525G
Specifications
| Ethyl Azidoacetate | |
| Yellow | |
| C4H7N3O2 | |
| 25 g | |
| ethyl azidoacetate, azidoacetic acid ethyl ester, acetic acid, azido-, ethyl ester, azido-acetic acid ethyl ester, ccris 3368, acetic acid, 2-azido-,ethyl ester, ethyl azido-acetate, ethyl2-azidoacetate, ethyl-2-azidoacetate, ethylazidoacetatesolution | |
| CCOC(=O)CN=[N+]=[N-] | |
| 129.119 | |
| 129.12 | |
| Liquid |
| 637-81-0 | |
| 164°C | |
| MFCD00190177 | |
| 1993 | |
| HVJJYOAPXBPQQV-UHFFFAOYSA-N | |
| ethyl 2-azidoacetate | |
| 69480 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 637-81-0 | |
| 129.119 | |
| HVJJYOAPXBPQQV-UHFFFAOYSA-N | |
| 69480 | |
| CCOC(=O)CN=[N+]=[N-] |
| C4H7N3O2 | |
| MFCD00190177 | |
| ethyl azidoacetate, azidoacetic acid ethyl ester, acetic acid, azido-, ethyl ester, azido-acetic acid ethyl ester, ccris 3368, acetic acid, 2-azido-,ethyl ester, ethyl azido-acetate, ethyl2-azidoacetate, ethyl-2-azidoacetate, ethylazidoacetatesolution | |
| ethyl 2-azidoacetate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator