missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Arachidate 95.0+%, TCI America™
$369.08 - $924.57
Chemical Identifiers
| CAS | 18281-05-5 |
|---|---|
| Molecular Formula | C22H44O2 |
| Molecular Weight (g/mol) | 340.59 |
| MDL Number | MFCD00056233 |
| InChI Key | YBKSMWBLSBAFBQ-UHFFFAOYSA-N |
| Synonym | Icosanoic Acid Ethyl Ester, Arachidic Acid Ethyl Ester, Ethyl Icosanoate |
| PubChem CID | 29009 |
| ChEBI | CHEBI:84859 |
| IUPAC Name | ethyl icosanoate |
| SMILES | CCCCCCCCCCCCCCCCCCCC(=O)OCC |
Chemical Identifiers
| 18281-05-5 | |
| 340.59 | |
| YBKSMWBLSBAFBQ-UHFFFAOYSA-N | |
| 29009 | |
| ethyl icosanoate |
| C22H44O2 | |
| MFCD00056233 | |
| Icosanoic Acid Ethyl Ester, Arachidic Acid Ethyl Ester, Ethyl Icosanoate | |
| CHEBI:84859 | |
| CCCCCCCCCCCCCCCCCCCC(=O)OCC |
Specifications
| 18281-05-5 | |
| White-Yellow | |
| C22H44O2 | |
| 5 g | |
| YBKSMWBLSBAFBQ-UHFFFAOYSA-N | |
| ethyl icosanoate | |
| 29009 | |
| 340.59 | |
| Crystalline Powder |
| 44°C | |
| 215°C | |
| MFCD00056233 | |
| Icosanoic Acid Ethyl Ester, Arachidic Acid Ethyl Ester, Ethyl Icosanoate | |
| CCCCCCCCCCCCCCCCCCCC(=O)OCC | |
| 340.59 | |
| CHEBI:84859 | |
| ≥95.0% (GC) | |
| Ethyl Arachidate |
Safety and Handling
EINECSNumber : (2)-0798
TSCA : No