accessibility menu, dialog, popup

UserName

CAS RN 18281-05-5

H3COOCH3

CAS RN 18281-05-5

IUPAC Name: ethyl icosanoate
Synonyms: ethyl eicosanoate
Molecular Weight (g/mol): 340.59
Molecular Formula: C22H44O2
InChi Key: YBKSMWBLSBAFBQ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCC(=O)OCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results