missing translation for 'onlineSavingsMsg'
Learn More

Ethyl alpha,4-dibromophenylacetate, 97+%

Catalog No. AAH2809306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2809306 5 g
AAH2809314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2809306 Supplier Thermo Scientific Chemicals Supplier No. H2809306
Only null left

CAS: 77143-76-1 | C10H10Br2O2 | 321.996 g/mol

It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 77143-76-1
Molecular Formula C10H10Br2O2
Molecular Weight (g/mol) 321.996
MDL Number MFCD09953497
InChI Key PPRAWWBOUOBAON-UHFFFAOYSA-N
Synonym ethyl 2-bromo-2-4-bromophenyl acetate, ethyl 2-bromo-4-bromophenyl acetate, ethyl bromo 4-bromophenyl acetate, alpha,4-dibromophenylacetic acid ethyl ester, ethyl alpha,4-dibromophenylacetate, ethyl2-bromo-2-4-bromophenyl acetate, ethyl 2-bromanyl-2-4-bromophenyl ethanoate, benzeneacetic acid, ?,4-dibromo-, ethyl ester, benzeneacetic acid, a,4-dibromo-, ethyl ester
PubChem CID 12754876
IUPAC Name ethyl 2-bromo-2-(4-bromophenyl)acetate
SMILES CCOC(=O)C(C1=CC=C(C=C1)Br)Br

Specifications

Density 1.642
Boiling Point 140°C (5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.571
Quantity 5 g
UN Number UN3265
Solubility Information Slightly soluble in water.
Formula Weight 321.99
Percent Purity ≥97%
Chemical Name or Material Ethyl alpha,4-dibromophenylacetate
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only