missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol, for spectroscopy, anhydrous, denat. with 5% 2-Propanol + 5% Methanol
CAS: 64-17-5 | C2H6O | 46.069 g/mol
Supplier: Thermo Scientific Chemicals 224090010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for UV/VIS spectroscopySpecifications
| Ethanol | |
| 0.01 max. at 300nm, 0.04 max. at 250nm, 0.65 max. at 210nm, 1 max. at 205nm | |
| 1 L | |
| Liquid | |
| C2H6O | |
| Vapor Pressure: 59mbar at 20°C | |
| 14, 3761 | |
| LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
| ethanol | |
| 702 | |
| 46.06 | |
| Spectroscopy |
| 64-17-5,67-56-1,67-63-0 | |
| 1.59 | |
| Glass Bottle | |
| 0.0005% max. | |
| C2H5OH | |
| 0.1% Max. (Coulometric) | |
| ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol | |
| CCO | |
| 46.069 | |
| CHEBI:16236 | |
| 4.0 to 6.0% (2-propanol) |
Chemical Identifiers
| 64-17-5 | |
| 46.069 | |
| ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol | |
| CHEBI:16236 | |
| CCO |
| C2H6O | |
| LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
| 702 | |
| ethanol |
For Manufacturing and Laboratory Use. Not for beverage use