missing translation for 'onlineSavingsMsg'
Learn More

Ethyl acrylate, 99.5%, stabilized

Catalog No. p-3736419
Change view
Click to view available options
Quantity:
25 mL
1 L
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC117980250 25 mL Glass bottle
AC117980010 1 L Glass bottle
AC117980025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC117980250 Supplier Thermo Scientific Chemicals Supplier No. 117980250

CAS: 140-88-5 | C5H8O2 | 100.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 140-88-5 , 150-76-5
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00009188
InChI Key JIGUQPWFLRLWPJ-UHFFFAOYSA-N
Synonym ethyl acrylate, acrylic acid ethyl ester, ethyl propenoate, 2-propenoic acid, ethyl ester, ethyl 2-propenoate, ethylacrylaat, ethylakrylat, etil acrilato, acrylic acid, ethyl ester, aethylacrylat
PubChem CID 8821
ChEBI CHEBI:82327
IUPAC Name ethyl prop-2-enoate
SMILES CCOC(=O)C=C

Specifications

Melting Point -71°C
Density 0.9200g/mL
Boiling Point 99°C
Flash Point 16°C
Infrared Spectrum Authentic
Assay Percent Range 99.40% min. (GC)
Packaging Glass bottle
Refractive Index 1.4050 to 1.407
Linear Formula H2C=CHCO2C2H5
Quantity 25 mL
Beilstein 02, 399
Fieser 06,251; 13,272
Specific Gravity 0.92
Merck Index 15, 3813
Solubility Information Solubility in water: 1.5% (25°C). Other solubilities: freely miscible with most organic solvents
Stabilizer 10 to 20ppm MEHQ
Viscosity 0.58 mPa.s (20°C)
Formula Weight 100.12
Percent Purity 99.50%
Physical Form Liquid
Chemical Name or Material Ethyl acrylate, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Toxic if inhaled
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Ground/Bond container and
EINECSNumber 205-438-8

RUO – Research Use Only