Learn More
Ethyl acrylate, 99.5%, stabilized
CAS: 140-88-5 | C5H8O2 | 100.12 g/mol
Supplier: Thermo Scientific Chemicals 117980025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Ethyl acrylate | |
| 140-88-5,150-76-5 | |
| 0.9200g/mL | |
| 16°C | |
| 99.40% min. (GC) | |
| C5H8O2 | |
| H2C=CHCO2C2H5 | |
| 2.5 L | |
| 06,251; 13,272 | |
| 15, 3813 | |
| Solubility in water: 1.5% (25°C). Other solubilities: freely miscible with most organic solvents | |
| CCOC(=O)C=C | |
| 100.12 | |
| 8821 | |
| CHEBI:82327 | |
| 99.50% |
| Stabilized | |
| -71°C | |
| 99°C | |
| Authentic | |
| Glass bottle | |
| 1.4050 to 1.407 | |
| MFCD00009188 | |
| 02, 399 | |
| 0.92 | |
| ethyl acrylate, acrylic acid ethyl ester, ethyl propenoate, 2-propenoic acid, ethyl ester, ethyl 2-propenoate, ethylacrylaat, ethylakrylat, etil acrilato, acrylic acid, ethyl ester, aethylacrylat | |
| JIGUQPWFLRLWPJ-UHFFFAOYSA-N | |
| ethyl prop-2-enoate | |
| 10 to 20ppm MEHQ | |
| 0.58 mPa.s (20°C) | |
| 100.12 | |
| Liquid |
Chemical Identifiers
| 140-88-5 | |
| 100.12 | |
| JIGUQPWFLRLWPJ-UHFFFAOYSA-N | |
| 8821 | |
| ethyl prop-2-enoate |
| C5H8O2 | |
| MFCD00009188 | |
| ethyl acrylate, acrylic acid ethyl ester, ethyl propenoate, 2-propenoic acid, ethyl ester, ethyl 2-propenoate, ethylacrylaat, ethylakrylat, etil acrilato, acrylic acid, ethyl ester, aethylacrylat | |
| CHEBI:82327 | |
| CCOC(=O)C=C |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Toxic if inhaled
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Ground/Bond container and
GHS Signal Word: Danger
EINECSNumber : 205-438-8
RUO – Research Use Only