missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetohydroximate 95.0+%, TCI America™

Catalog No. p-7117378
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13395G 5 g
A133925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A13395G Supplier TCI America Supplier No. A13395G
Only null left

Chemical Identifiers

CAS 10576-12-2
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.12
MDL Number MFCD00002114,MFCD00002114
InChI Key QWKAVVNRCKPKNM-SNAWJCMRSA-N
Synonym ethyl n-hydroxyacetimidate, n-hydroxyethanimidic acid, ethyl ester, ethyl acetohydroximate, ethyl n-hydroxyethanimidate, e-ethyl n-hydroxyethanimidate, e-ethyl n-hydroxyacetimidate, ethyl 1e-n-hydroxyethanimidate, snx`lfelmmpqh, ethyl acethydroxamate, 1-ethoxyethanone oxime
PubChem CID 6386647
IUPAC Name (E)-(ethyl N-hydroxyethanimidate)
SMILES CCO\C(C)=N\O

Specifications

Color White-Yellow
Boiling Point 60°C
Quantity 5 g
Formula Weight 103.12
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Ethyl Acetohydroximate
TSCA Yes
Recommended Storage Refrigerator