missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC), Honeywell Riedel-de Haën™
Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.5% (GC)
Supplier: Honeywell Chemicals 33211180KG
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
| Ethyl acetate | |
| 141-78-6 | |
| 77°C | |
| 2.2 to 11.5% (38°F) | |
| 801°F | |
| Metal Drum | |
| C4H8O2 | |
| MFCD00009171 | |
| UN1173 | |
| ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester | |
| XEKOWRVHYACXOJ-UHFFFAOYSA-N | |
| ethyl acetate | |
| 8857 | |
| 88.11g/mol | |
| ACS/Reagent ISO/Reagent Ph. Eur./puriss. p.a. |
| -84°C | |
| 0.902g/mL at 25°C | |
| 180 kg | |
| −3°C (26.6°F) | |
| Like fruit | |
| Liquid | |
| CH3COOC2H5 | |
| 73mmHg (20°C) | |
| 506104 | |
| ∽90g/L | |
| CCOC(=O)C | |
| 88.106 | |
| CHEBI:27750 | |
| ≥99.5% (GC) |
Chemical Identifiers
| 141-78-6 | |
| 88.106 | |
| XEKOWRVHYACXOJ-UHFFFAOYSA-N | |
| 8857 | |
| ethyl acetate |
| C4H8O2 | |
| MFCD00009171 | |
| ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester | |
| CHEBI:27750 | |
| CCOC(=O)C |
Safety and Handling
P210-P305 + P351 + P338-P370 + P378-P403 + P235
Danger
DOTInformation : Class 3 - PG 2 - Ethyl acetate
RTECSNumber : 205-500-4