missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC), Honeywell Riedel-de Haën™
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Specifications
| Melting Point | -84°C |
| Density | 0.902g/mL at 25°C |
| Boiling Point | 77°C |
| Quantity | 1 L |
| Explosion Limit | 2.2 to 11.5% (38°F) |
| Flash Point | −3°C (26.6°F) |
| Ignition Point | 801°F |
| Odor | Like fruit |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | P210-P305 + P351 + P338-P370 + P378-P403 + P235 |
| DOTInformation | Class 3 - PG 2 - Ethyl acetate |
| RTECSNumber | 205-500-4 |