missing translation for 'onlineSavingsMsg'
Learn More

Ethyl acetate, Analytical Standard, ≥99.9% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019346
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019346 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019346 Supplier Merck Emd Millipore Supplier No. 589585ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ethyl acetate may be used as an extractant solvent in the analysis of moxalactum in serum samples using high performance liquid chromatography (HPLC).

Specifications

CAS 141-78-6
Vapor Pressure 73 mmHg (20°C)
Vapor Density 3 (20°C, vs air)
Density 0.902 g/mL (at 25°C)
Physical Form Neat
Linear Formula CH3COOC2H5
MDL Number MFCD00009171
Melting Point -84°C
Boiling Point 76.5°C to 77.5°C
Synonym EtOAc
Solubility Soluble in water
Quantity 5 mL
Formula Weight 88.11
Percent Purity ≥99.9% (GC)
Grade Analytical Standard
Show More Show Less