Learn More
Ethyl acetamidocyanoacetate, 98%, Thermo Scientific Chemicals
$129.40 - $129.40
Chemical Identifiers
| CAS | 4977-62-2 |
|---|---|
| Molecular Formula | C7H10N2O3 |
| Molecular Weight (g/mol) | 170.17 |
| MDL Number | MFCD00009133 |
| InChI Key | SLIRLABNGAZSHX-UHFFFAOYNA-N |
| Synonym | ethyl acetamidocyanoacetate, ethyl acetaminocyanoacetate, n-acetyl-2-cyanoglycine ethyl ester, ethyl 2-cyano-2-acetamidoacetate, acetic acid, acetylamino cyano-, ethyl ester, ethyl 2-acetylamino-2-cyanoacetate, ethyl acetamido cyano acetate, glycine, n-acetyl-2-cyano-, ethyl ester, acmc-209ki4, ethyl acetamido-cyanoacetate |
| PubChem CID | 95422 |
| IUPAC Name | ethyl 2-acetamido-2-cyanoacetate |
| SMILES | CCOC(=O)C(NC(C)=O)C#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0767409
|
Thermo Scientific Chemicals
L0767409 |
10 g |
Each for $129.40
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 4977-62-2 | |
| 170.17 | |
| SLIRLABNGAZSHX-UHFFFAOYNA-N | |
| 95422 | |
| CCOC(=O)C(NC(C)=O)C#N |
| C7H10N2O3 | |
| MFCD00009133 | |
| ethyl acetamidocyanoacetate, ethyl acetaminocyanoacetate, n-acetyl-2-cyanoglycine ethyl ester, ethyl 2-cyano-2-acetamidoacetate, acetic acid, acetylamino cyano-, ethyl ester, ethyl 2-acetylamino-2-cyanoacetate, ethyl acetamido cyano acetate, glycine, n-acetyl-2-cyano-, ethyl ester, acmc-209ki4, ethyl acetamido-cyanoacetate | |
| ethyl 2-acetamido-2-cyanoacetate |
Specifications
| 4977-62-2 | |
| C7H10N2O3 | |
| 10 g | |
| ethyl acetamidocyanoacetate, ethyl acetaminocyanoacetate, n-acetyl-2-cyanoglycine ethyl ester, ethyl 2-cyano-2-acetamidoacetate, acetic acid, acetylamino cyano-, ethyl ester, ethyl 2-acetylamino-2-cyanoacetate, ethyl acetamido cyano acetate, glycine, n-acetyl-2-cyano-, ethyl ester, acmc-209ki4, ethyl acetamido-cyanoacetate | |
| CCOC(=O)C(NC(C)=O)C#N | |
| 170.17 | |
| 170.17 | |
| Ethyl acetamidocyanoacetate |
| 128°C to 130°C | |
| MFCD00009133 | |
| 879845 | |
| SLIRLABNGAZSHX-UHFFFAOYNA-N | |
| ethyl 2-acetamido-2-cyanoacetate | |
| 95422 | |
| 98% |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 225-624-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only