accessibility menu, dialog, popup

UserName

CAS RN 4977-62-2

IUPAC Name: ethyl 2-cyano-2-acetamidoacetate
Synonyms: ethyl 2-cyano-2-acetamidoacetate
Molecular Weight (g/mol): 170.17
Molecular Formula: C7H10N2O3
InChi Key: SLIRLABNGAZSHX-UHFFFAOYNA-N
SMILES: CCOC(=O)C(NC(C)=O)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results