missing translation for 'onlineSavingsMsg'
Learn More

Ethyl acetamidocyanoacetate, 98%, Thermo Scientific Chemicals

Catalog No. AAL0767409
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0767409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0767409 Supplier Thermo Scientific Chemicals Supplier No. L0767409
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4977-62-2 | Molecular Formula: C7H10N2O3 | Molecular Weight: 170.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4977-62-2
Molecular Formula C7H10N2O3
Molecular Weight (g/mol) 170.17
MDL Number MFCD00009133
InChI Key SLIRLABNGAZSHX-UHFFFAOYNA-N
Synonym ethyl acetamidocyanoacetate, ethyl acetaminocyanoacetate, n-acetyl-2-cyanoglycine ethyl ester, ethyl 2-cyano-2-acetamidoacetate, acetic acid, acetylamino cyano-, ethyl ester, ethyl 2-acetylamino-2-cyanoacetate, ethyl acetamido cyano acetate, glycine, n-acetyl-2-cyano-, ethyl ester, acmc-209ki4, ethyl acetamido-cyanoacetate
PubChem CID 95422
IUPAC Name ethyl 2-acetamido-2-cyanoacetate
SMILES CCOC(=O)C(NC(C)=O)C#N

Specifications

Melting Point 128°C to 130°C
Quantity 10 g
Beilstein 879845
Formula Weight 170.17
Percent Purity 98%
Chemical Name or Material Ethyl acetamidocyanoacetate
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 225-624-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only