missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Abietate 80.0+%, TCI America™

Catalog No. A000225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A000225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A000225G Supplier TCI America Supplier No. A000225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 631-71-0
Molecular Formula C22H34O2
Molecular Weight (g/mol) 330.51
MDL Number MFCD00028860
InChI Key AGUBCDYYAKENKG-YVNJGZBMSA-N
Synonym Abietic Acid Ethyl Ester, Ethyl Rosinate, Rosin Acid Ethyl Ester
PubChem CID 53642177
IUPAC Name ethyl (1R,4aR,4bR,10aR)-1,4a-dimethyl-7-(propan-2-yl)-1,2,3,4,4a,4b,5,6,10,10a-decahydrophenanthrene-1-carboxylate
SMILES CCOC(=O)[C@]1(C)CCC[C@]2(C)[C@H]3CCC(=CC3=CC[C@@H]12)C(C)C

Specifications

Color Yellow
Boiling Point 350°C
Quantity 25 g
Formula Weight 330.51
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Abietate