missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 7-Hydroxycoumarin-3-carboxylate 98.0+%, TCI America™

Catalog No. E1017200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E1017200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E1017200MG Supplier TCI America Supplier No. E1017200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6093-71-6
Molecular Formula C12H10O5
Molecular Weight (g/mol) 234.207
MDL Number MFCD00017641
InChI Key IETDBZQIWIJQJG-UHFFFAOYSA-N
Synonym 7-Hydroxycoumarin-3-carboxylic Acid Ethyl Ester, Ethyl Umbelliferone-3-carboxylate, Umbelliferone-3-carboxylic Acid Ethyl Ester
PubChem CID 5289613
IUPAC Name ethyl 7-hydroxy-2-oxochromene-3-carboxylate
SMILES CCOC(=O)C1=CC2=C(C=C(C=C2)O)OC1=O

Specifications

Melting Point 172°C
Quantity 200 mg
Formula Weight 234.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 7-Hydroxycoumarin-3-carboxylate
TSCA No
Recommended Storage Freezer