accessibility menu, dialog, popup

UserName

CAS RN 6093-71-6

IUPAC Name: ethyl 7-hydroxy-2-oxo-2H-chromene-3-carboxylate
Synonyms: ethyl 7-hydroxy-2-oxochromene-3-carboxylate
Molecular Weight (g/mol): 234.21
Molecular Formula: C12H10O5
InChi Key: IETDBZQIWIJQJG-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC2=CC=C(O)C=C2OC1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results