missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 7-(Diethylamino)coumarin-3-carboxylate 98.0+%, TCI America™
$141.67 - $423.14
Chemical Identifiers
| CAS | 28705-46-6 |
|---|---|
| Molecular Formula | C16H19NO4 |
| Molecular Weight (g/mol) | 289.331 |
| MDL Number | MFCD00227484 |
| InChI Key | MSOLGAJLRIINNF-UHFFFAOYSA-N |
| Synonym | 7-(Diethylamino)coumarin-3-carboxylic Acid Ethyl Ester |
| PubChem CID | 120014 |
| IUPAC Name | ethyl 7-(diethylamino)-2-oxochromene-3-carboxylate |
| SMILES | CCN(CC)C1=CC2=C(C=C1)C=C(C(=O)O2)C(=O)OCC |
Chemical Identifiers
| 28705-46-6 | |
| 289.331 | |
| MSOLGAJLRIINNF-UHFFFAOYSA-N | |
| 120014 | |
| CCN(CC)C1=CC2=C(C=C1)C=C(C(=O)O2)C(=O)OCC |
| C16H19NO4 | |
| MFCD00227484 | |
| 7-(Diethylamino)coumarin-3-carboxylic Acid Ethyl Ester | |
| ethyl 7-(diethylamino)-2-oxochromene-3-carboxylate |
Specifications
| 28705-46-6 | |
| C16H19NO4 | |
| 1 g | |
| MSOLGAJLRIINNF-UHFFFAOYSA-N | |
| ethyl 7-(diethylamino)-2-oxochromene-3-carboxylate | |
| 120014 | |
| ≥98.0% (GC,T) | |
| Ethyl 7-(Diethylamino)coumarin-3-carboxylate |
| 86°C | |
| MFCD00227484 | |
| 7-(Diethylamino)coumarin-3-carboxylic Acid Ethyl Ester | |
| CCN(CC)C1=CC2=C(C=C1)C=C(C(=O)O2)C(=O)OCC | |
| 289.331 | |
| 289.33 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes