missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 7-(Diethylamino)coumarin-3-carboxylate 98.0+%, TCI America™

Supplier:  TCI America E06421G

 View more versions of this product

Catalog No. E06421G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ethyl 7-(Diethylamino)coumarin-3-carboxylate
86°C
MFCD00227484
7-(Diethylamino)coumarin-3-carboxylic Acid Ethyl Ester
CCN(CC)C1=CC2=C(C=C1)C=C(C(=O)O2)C(=O)OCC
289.331
289.33
Crystalline Powder
28705-46-6
C16H19NO4
1 g
MSOLGAJLRIINNF-UHFFFAOYSA-N
ethyl 7-(diethylamino)-2-oxochromene-3-carboxylate
120014
≥98.0% (GC,T)

Chemical Identifiers

28705-46-6
289.331
MSOLGAJLRIINNF-UHFFFAOYSA-N
120014
CCN(CC)C1=CC2=C(C=C1)C=C(C(=O)O2)C(=O)OCC
C16H19NO4
MFCD00227484
7-(Diethylamino)coumarin-3-carboxylic Acid Ethyl Ester
ethyl 7-(diethylamino)-2-oxochromene-3-carboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions