missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 7-Chloro-2-oxoheptanoate 98.0+%, TCI America™

Catalog No. E08275G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08275G 5 g
E082725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E08275G Supplier TCI America Supplier No. E08275G
Only null left

Chemical Identifiers

CAS 78834-75-0
Molecular Formula C9H15ClO3
Molecular Weight (g/mol) 206.666
MDL Number MFCD07698675
InChI Key YJJLIIMRHGRCFM-UHFFFAOYSA-N
Synonym 7-Chloro-2-oxoheptanoic Acid Ethyl Ester
PubChem CID 3018793
IUPAC Name ethyl 7-chloro-2-oxoheptanoate
SMILES CCOC(=O)C(=O)CCCCCCl

Specifications

Color Yellow
Quantity 5 g
Formula Weight 206.67
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 7-Chloro-2-oxoheptanoate
TSCA No
Recommended Storage Refrigerator