accessibility menu, dialog, popup

UserName

H3COOOCl

CAS RN 78834-75-0

IUPAC Name: ethyl 7-chloro-2-oxoheptanoate
Synonyms: ethyl 7-chloro-2-oxoheptanoate
Molecular Weight (g/mol): 206.67
Molecular Formula: C9H15ClO3
InChi Key: YJJLIIMRHGRCFM-UHFFFAOYSA-N
SMILES: CCOC(=O)C(=O)CCCCCCl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results