missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 7-Chloro-1-(2,4-difluorophenyl)-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate 98.0+%, TCI America™

Catalog No. p-7129381
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
E11805G 5 g
E118025G 25 g
2 options

Catalog No. E11805G

Supplier: TCI America E11805G

Only null left

Chemical Identifiers

CAS 100491-29-0
Molecular Formula C17H10ClF3N2O3
Molecular Weight (g/mol) 382.723
MDL Number MFCD01863285
InChI Key JLSXYCZRMYCIMY-UHFFFAOYSA-N
Synonym ethyl 7-chloro-1-2,4-difluorophenyl-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate, ethyl 7-chloro-1-2,4-difluorophenyl-6-fluoro-4-oxo-1,8-naphthyridine-3-carboxylate, 7-chloro-1-2,4-difluorophenyl-6-fluoro-1,4-dihydro-4-oxo-1,8-naphthyridine-3-carboxylic acid ethyl ester, 7-chloro-1-2,4-difluoro-phenyl-6-fluoro-4-oxo-1,4-dihydro-1,8 naphthyridine-3-carboxylic acid ethyl ester, ethyl 1-2,4-difluorophenyl-7-cl-6-f-4-o-hydropyridino 2,3-b pyridine-3-carboxylate, 7-chloro-6-fluoro-1-2,4-difluorophenyl-1,4-dihydro-4-oxo-1,8-naphthyridine-3-carboxylic acid ethyl ester, ethyl 1-2,4-difluorophenyl-7-chloro-6-fluoro-1,4-dihydro-4-oxo-1,8-naphthyridine-3-carboxylate, ethyl 1-2,4-difluorophenyl-7-chloro-6-fluoro-4-oxohydropyridino 2,3-b pyridi ne-3-carboxylate, ethyl 1-2,4-difluorophenyl-7-chloro-6-fluoro-4-oxopyridino 2,3-b pyridine-3-carboxylate, ethyl 7-chloro-1-2,4-difluorophenyl-6-fluoro-1,4-dihydro-4-oxo-1,8-naphthyridine-3-carboxylate
PubChem CID 1268243
IUPAC Name ethyl 7-chloro-1-(2,4-difluorophenyl)-6-fluoro-4-oxo-1,8-naphthyridine-3-carboxylate
SMILES CCOC(=O)C1=CN(C2=NC(=C(C=C2C1=O)F)Cl)C3=C(C=C(C=C3)F)F

Specifications

Melting Point 215°C
Color White-Yellow
Quantity 5 g
UN Number 3077
Formula Weight 382.72
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 7-Chloro-1-(2,4-difluorophenyl)-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate
EINECSNumber (5)-6439
TSCA No