accessibility menu, dialog, popup

UserName

CAS RN 100491-29-0

IUPAC Name: ethyl 7-chloro-1-(2,4-difluorophenyl)-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate
Synonyms: ethyl 7-chloro-1-(2,4-difluorophenyl)-6-fluoro-4-oxo-1,8-naphthyridine-3-carboxylate
Molecular Weight (g/mol): 382.72
Molecular Formula: C17H10ClF3N2O3
InChi Key: JLSXYCZRMYCIMY-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN(C2=CC=C(F)C=C2F)C2=NC(Cl)=C(F)C=C2C1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results