missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 7-Chloro-1-(2,4-difluorophenyl)-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 215°C |
| Color | White-Yellow |
| Quantity | 25 g |
| UN Number | 3077 |
| Formula Weight | 382.72 |
| Percent Purity | ≥98.0% (HPLC,N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Ethyl 7-Chloro-1-(2,4-difluorophenyl)-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate |
Safety and Handling
| EINECSNumber | (5)-6439 |
| TSCA | No |