missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 7-Bromoheptanoate 97.0+%, TCI America™

Catalog No. B18805G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18805G 5 g
B188025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18805G Supplier TCI America Supplier No. B18805G

Chemical Identifiers

CAS 29823-18-5
Molecular Formula C9H17BrO2
Molecular Weight (g/mol) 237.14
MDL Number MFCD00037103
InChI Key OOBFNDGMAGSNKA-UHFFFAOYSA-N
PubChem CID 122490
IUPAC Name ethyl 7-bromoheptanoate
SMILES CCOC(=O)CCCCCCBr

Specifications

Color Yellow
Boiling Point 145°C
Quantity 5 g
Synonym 7-bromoheptanoic acid ethyl ester, ethyl-7-bromoheptanoate, heptanoic acid, 7-bromo-, ethyl ester, ethyl7-bromoheptanoate, br ch2 6cooet, ethyl 7-bromo-heptanoate, ethyl 7-bromanylheptanoate, acmc-209hb9, ethyl 7-bromoheptanoate, oobfndgmagsnka-uhfffaoysa
Formula Weight 237.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 7-Bromoheptanoate