missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 6-Methyl-2-oxo-4-phenyl-1,2,3,4-tetrahydropyrimidine-5-carboxylate 98.0+%, TCI America™

Catalog No. p-7129348
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E1131200MG 200 mg
E11311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E1131200MG Supplier TCI America Supplier No. E1131200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5395-36-8
Molecular Formula C14H16N2O3
Molecular Weight (g/mol) 260.293
MDL Number MFCD00124778
InChI Key URMNHHAUVFEMIG-UHFFFAOYSA-N
Synonym 6-Methyl-2-oxo-4-phenyl-1,2,3,4-tetrahydropyrimidine-5-carboxylic Acid Ethyl Ester
PubChem CID 220272
IUPAC Name ethyl 6-methyl-2-oxo-4-phenyl-3,4-dihydro-1H-pyrimidine-5-carboxylate
SMILES CCOC(=O)C1=C(NC(=O)NC1C2=CC=CC=C2)C

Specifications

Melting Point 208°C
Color White
Quantity 200 mg
Formula Weight 260.29
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 6-Methyl-2-oxo-4-phenyl-1,2,3,4-tetrahydropyrimidine-5-carboxylate