accessibility menu, dialog, popup

UserName

CAS RN 5395-36-8

IUPAC Name: ethyl 6-methyl-2-oxo-4-phenyl-1,2,3,4-tetrahydropyrimidine-5-carboxylate
Synonyms: ethyl 4-methyl-2-oxo-6-phenyl-3,6-dihydro-1H-pyrimidine-5-carboxylate
Molecular Weight (g/mol): 260.29
Molecular Formula: C14H16N2O3
InChi Key: URMNHHAUVFEMIG-UHFFFAOYNA-N
SMILES: CCOC(=O)C1=C(C)NC(=O)NC1C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results