missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 6-Methyl-2-oxo-3-cyclohexene-1-carboxylate (mixture of isomers) 95.0+%, TCI America™

Catalog No. e06465g
Change view
Click to view available options
Quantity:
5 g
Catalog No. Quantity
E06465G 5 g
1 options

Catalog No. E06465G

Supplier: TCI America E06465G

Only null left

Chemical Identifiers

CAS 3419-32-7
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.219
InChI Key CFQIYLYEYJYZEO-UHFFFAOYSA-N
Synonym Ethyl 3-Methyl-5-cyclohexen-1-one-2-carboxylate, 6-Methyl-2-oxo-3-cyclohexene-1-carboxylic Acid Ethyl Ester
PubChem CID 549414
IUPAC Name ethyl 6-methyl-2-oxocyclohex-3-ene-1-carboxylate
SMILES CCOC(=O)C1C(CC=CC1=O)C

Specifications

Color Yellow
Boiling Point 110°C
Quantity 5 g
Formula Weight 182.22
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 6-Methyl-2-oxo-3-cyclohexene-1-carboxylate (mixture of isomers)