accessibility menu, dialog, popup

UserName

H3COOOCH3

CAS RN 3419-32-7

IUPAC Name: ethyl 6-methyl-2-oxocyclohex-3-ene-1-carboxylate
Synonyms: ethyl 6-methyl-2-oxocyclohex-3-ene-1-carboxylate
Molecular Weight (g/mol): 182.22
Molecular Formula: C10H14O3
InChi Key: CFQIYLYEYJYZEO-UHFFFAOYNA-N
SMILES: CCOC(=O)C1C(C)CC=CC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results