missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 6-chloropyridine-2-carboxylate, 97%

Catalog No. AAH6170106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6170106 5 g
AAH6170103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6170106 Supplier Thermo Scientific Chemicals Supplier No. H6170106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 21190-89-6 | C8H8ClNO2 | 185.607 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21190-89-6
Molecular Formula C8H8ClNO2
Molecular Weight (g/mol) 185.607
MDL Number MFCD09878438
InChI Key ORSVWYSFUABOQU-UHFFFAOYSA-N
Synonym ethyl 6-chloropicolinate, ethyl 6-chloro-2-pyridinecarboxylate, 6-chloropyridine-2-carboxylic acid ethyl ester, 6-chloro-pyridine-2-carboxylic acid ethyl ester, 2-pyridinecarboxylicacid, 6-chloro-, ethyl ester, 2-pyridinecarboxylic acid, 6-chloro-, ethyl ester, pubchem17513, acmc-209fib, 2-chloro-6-ethoxycarbonyl-pyridine
PubChem CID 11063114
IUPAC Name ethyl 6-chloropyridine-2-carboxylate
SMILES CCOC(=O)C1=NC(=CC=C1)Cl

Specifications

Color Yellow
Quantity 5 g
Formula Weight 185.61
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material Ethyl 6-chloropyridine-2-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only