CAS RN 21190-89-6
IUPAC Name:
ethyl 6-chloropyridine-2-carboxylate
Synonyms:
ethyl 6-chloropyridine-2-carboxylate
Molecular Weight (g/mol):
185.61
Molecular Formula:
C8H8ClNO2
InChi Key:
ORSVWYSFUABOQU-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=NC(Cl)=CC=C1
1
–
1
of
1
results
1
Promotions Available
Ethyl 6-chloropyridine-2-carboxylate, 97%
Ethyl 6-chloropyridine-2-carboxylate, 97%
CAS: 21190-89-6 Molecular Formula: C8H8ClNO2 Molecular Weight (g/mol): 185.607 MDL Number: MFCD09878438 InChI Key: ORSVWYSFUABOQU-UHFFFAOYSA-N Synonym: ethyl 6-chloropicolinate,ethyl 6-chloro-2-pyridinecarboxylate,6-chloropyridine-2-carboxylic acid ethyl ester,6-chloro-pyridine-2-carboxylic acid ethyl ester,2-pyridinecarboxylicacid, 6-chloro-, ethyl ester,2-pyridinecarboxylic acid, 6-chloro-, ethyl ester,pubchem17513,acmc-209fib,2-chloro-6-ethoxycarbonyl-pyridine PubChem CID: 11063114 IUPAC Name: ethyl 6-chloropyridine-2-carboxylate SMILES: CCOC(=O)C1=NC(=CC=C1)Cl