missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 6-Chloronicotinate 97.0+%, TCI America™
$109.74 - $348.46
Chemical Identifiers
| CAS | 49608-01-7 |
|---|---|
| Molecular Formula | C8H8ClNO2 |
| Molecular Weight (g/mol) | 185.61 |
| MDL Number | MFCD00082739 |
| InChI Key | ILDJJTQWIZLGPO-UHFFFAOYSA-N |
| Synonym | ethyl 6-chloronicotinate, ethyl 6-chloro-3-pyridinecarboxylate, ethyl6-chloronicotinate, 6-chloro-nicotinic acid ethyl ester, 6-chloronicotinic acid ethyl ester, ethyl 6-chloro nicotinate, ethyl 2-chloro-5-pyridinecarboxylate, 3-pyridinecarboxylic acid, 6-chloro-, ethyl ester, pubchem1415, pubchem15179 |
| PubChem CID | 2799611 |
| IUPAC Name | ethyl 6-chloropyridine-3-carboxylate |
| SMILES | CCOC(=O)C1=CC=C(Cl)N=C1 |
Chemical Identifiers
| 49608-01-7 | |
| 185.61 | |
| ILDJJTQWIZLGPO-UHFFFAOYSA-N | |
| 2799611 | |
| CCOC(=O)C1=CC=C(Cl)N=C1 |
| C8H8ClNO2 | |
| MFCD00082739 | |
| ethyl 6-chloronicotinate, ethyl 6-chloro-3-pyridinecarboxylate, ethyl6-chloronicotinate, 6-chloro-nicotinic acid ethyl ester, 6-chloronicotinic acid ethyl ester, ethyl 6-chloro nicotinate, ethyl 2-chloro-5-pyridinecarboxylate, 3-pyridinecarboxylic acid, 6-chloro-, ethyl ester, pubchem1415, pubchem15179 | |
| ethyl 6-chloropyridine-3-carboxylate |
Specifications
| 49608-01-7 | |
| 150°C | |
| MFCD00082739 | |
| ethyl 6-chloronicotinate, ethyl 6-chloro-3-pyridinecarboxylate, ethyl6-chloronicotinate, 6-chloro-nicotinic acid ethyl ester, 6-chloronicotinic acid ethyl ester, ethyl 6-chloro nicotinate, ethyl 2-chloro-5-pyridinecarboxylate, 3-pyridinecarboxylic acid, 6-chloro-, ethyl ester, pubchem1415, pubchem15179 | |
| CCOC(=O)C1=CC=C(Cl)N=C1 | |
| 185.61 | |
| 185.61 | |
| Crystalline Lumps |
| White-Yellow | |
| C8H8ClNO2 | |
| 5 g | |
| ILDJJTQWIZLGPO-UHFFFAOYSA-N | |
| ethyl 6-chloropyridine-3-carboxylate | |
| 2799611 | |
| ≥97.0% (GC) | |
| Ethyl 6-Chloronicotinate |
Safety and Handling
TSCA : No