missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 6-Chloronicotinate 97.0+%, TCI America™
Supplier: TCI America E085225G
Specifications
| Ethyl 6-Chloronicotinate | |
| White-Yellow | |
| C8H8ClNO2 | |
| 25 g | |
| ILDJJTQWIZLGPO-UHFFFAOYSA-N | |
| ethyl 6-chloropyridine-3-carboxylate | |
| 2799611 | |
| ≥97.0% (GC) |
| 49608-01-7 | |
| 150°C | |
| MFCD00082739 | |
| ethyl 6-chloronicotinate, ethyl 6-chloro-3-pyridinecarboxylate, ethyl6-chloronicotinate, 6-chloro-nicotinic acid ethyl ester, 6-chloronicotinic acid ethyl ester, ethyl 6-chloro nicotinate, ethyl 2-chloro-5-pyridinecarboxylate, 3-pyridinecarboxylic acid, 6-chloro-, ethyl ester, pubchem1415, pubchem15179 | |
| CCOC(=O)C1=CC=C(Cl)N=C1 | |
| 185.61 | |
| 185.61 | |
| Crystalline Lumps |
Chemical Identifiers
| 49608-01-7 | |
| 185.61 | |
| ILDJJTQWIZLGPO-UHFFFAOYSA-N | |
| 2799611 | |
| CCOC(=O)C1=CC=C(Cl)N=C1 |
| C8H8ClNO2 | |
| MFCD00082739 | |
| ethyl 6-chloronicotinate, ethyl 6-chloro-3-pyridinecarboxylate, ethyl6-chloronicotinate, 6-chloro-nicotinic acid ethyl ester, 6-chloronicotinic acid ethyl ester, ethyl 6-chloro nicotinate, ethyl 2-chloro-5-pyridinecarboxylate, 3-pyridinecarboxylic acid, 6-chloro-, ethyl ester, pubchem1415, pubchem15179 | |
| ethyl 6-chloropyridine-3-carboxylate |
Safety and Handling
TSCA : No