missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 6-Bromohexanoate 98.0+%, TCI America™

Catalog No. B146525G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B146525G 25 g
B1465250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B146525G Supplier TCI America Supplier No. B146525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 25542-62-5
Molecular Formula C8H15BrO2
Molecular Weight (g/mol) 223.11
MDL Number MFCD00000270
InChI Key DXBULVYHTICWKT-UHFFFAOYSA-N
Synonym 6-bromohexanoic acid ethyl ester, ethyl 6-bromocaproate, ethyl 6-bromocapronate, 6-bromocaproic acid ethyl ester, ethyl omega-bromocaproate, hexanoic acid, 6-bromo-, ethyl ester, ethyl .omega.-bromocaproate, 6-bromo-hexanoic acid ethyl ester, ethyl 6bromohexanoate, pubchem3953
PubChem CID 117544
IUPAC Name ethyl 6-bromohexanoate
SMILES CCOC(=O)CCCCCBr

Specifications

Color Yellow
Boiling Point 130°C
Quantity 25 g
UN Number 3272
Formula Weight 223.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 6-Bromohexanoate